Anti-NEDD9 Mouse Monoclonal Antibody [clone: 584825]

Supplier: R & D Systems
MAB5459
102886-026EA 566.89 USD
102886-026
Anti-NEDD9 Mouse Monoclonal Antibody [clone: 584825]
Antibodies
Type: Primary
Antigen Symbol: NEDD9
Antigen Name: neural precursor cell expressed, developmentally down-regulated 9
Antigen Synonyms: CAS-L, CAS2, CASL, CASS2, HEF1
Clonality: Monoclonal
Clone: 584825
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR