Anti-GPX4 Mouse Monoclonal Antibody [clone: 565320]

Supplier: R & D Systems
MAB5457
220021-179EA 616.06 USD
220021-179
Anti-GPX4 Mouse Monoclonal Antibody [clone: 565320]
Antibodies
Type: Primary
Antigen Symbol: GPX4
Antigen Name: glutathione peroxidase 4
Antigen Synonyms: GPx-4, GSHPx-4, PHGPx, mtPHGPx, snGPx
Clonality: Monoclonal
Clone: 565320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR