Anti-IL5 Mouse Monoclonal Antibody [clone: 299257]

Supplier: R & D Systems
MAB5451
220021-177EA 561.11 USD
220021-177
Anti-IL5 Mouse Monoclonal Antibody [clone: 299257]
Antibodies
Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone: 299257
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR