Anti-IL5 Mouse Monoclonal Antibody [clone: 119905]

Supplier: R & D Systems
MAB545
220021-176EA 763.57 USD
220021-176
Anti-IL5 Mouse Monoclonal Antibody [clone: 119905]
Antibodies
Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone: 119905
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR