Anti-CD44 Mouse Monoclonal Antibody [clone: 69-S5]

Supplier: R & D Systems
MAB5449
220021-175EA 561.11 USD
220021-175
Anti-CD44 Mouse Monoclonal Antibody [clone: 69-S5]
Antibodies
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 69-S5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR