Anti-CLC Mouse Monoclonal Antibody [clone: 561603]

Supplier: R & D Systems
MAB5447
102886-024EA 594.37 USD
102886-024
Anti-CLC Mouse Monoclonal Antibody [clone: 561603]
Antibodies
Type: Primary
Antigen: CLC
Clonality: Monoclonal
Clone: 561603
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR