Anti-LRFN4 Mouse Monoclonal Antibody [clone: 578102]

Supplier: R & D Systems
MAB5445
102886-022EA 577.02 USD
102886-022
Anti-LRFN4 Mouse Monoclonal Antibody [clone: 578102]
Antibodies
Type: Primary
Antigen: LRFN4
Clonality: Monoclonal
Clone: 578102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR