Anti-ADGRF4 Mouse Monoclonal Antibody [clone: 5273]

Supplier: R & D Systems
MAB5437
220021-174EA 577.02 USD
220021-174
Anti-ADGRF4 Mouse Monoclonal Antibody [clone: 5273]
Antibodies
Type: Primary
Antigen: ADGRF4
Clonality: Monoclonal
Clone: 5273
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR