Anti-CREB1 Rat Monoclonal Antibody [clone: 485422]

Supplier: R & D Systems
MAB5435
220021-172EA 594.37 USD
220021-172
Anti-CREB1 Rat Monoclonal Antibody [clone: 485422]
Antibodies
Type: Primary
Antigen: CREB1
Clonality: Monoclonal
Clone: 485422
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR