Anti-PCDH8 Mouse Monoclonal Antibody [clone: 562615]

Supplier: R & D Systems
MAB5434
102886-020EA 594.37 USD
102886-020
Anti-PCDH8 Mouse Monoclonal Antibody [clone: 562615]
Antibodies
Type: Primary
Antigen: PCDH8
Clonality: Monoclonal
Clone: 562615
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR