Anti-SERPINA9 Mouse Monoclonal Antibody [clone: 585302]

Supplier: R & D Systems
MAB5429
220021-169EA 584.25 USD
220021-169
Anti-SERPINA9 Mouse Monoclonal Antibody [clone: 585302]
Antibodies
Type: Primary
Antigen: SERPINA9
Clonality: Monoclonal
Clone: 585302
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR