Anti-TGM7 Rat Monoclonal Antibody [clone: 602708]

Supplier: R & D Systems
MAB5426
102886-018EA 561.11 USD
102886-018
Anti-TGM7 Rat Monoclonal Antibody [clone: 602708]
Antibodies
Type: Primary
Antigen: TGM7
Clonality: Monoclonal
Clone: 602708
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR