Anti-BoNT-C1 Mouse Monoclonal Antibody [clone: 580223]

Supplier: R & D Systems
MAB5425
102886-016EA 561.11 USD
102886-016
Anti-BoNT-C1 Mouse Monoclonal Antibody [clone: 580223]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: BoNT-C1 Heavy Chain
Antigen Synonyms:
Clonality: Monoclonal
Clone: 580223
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: C. botulinum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR