Anti-EPHB1 Mouse Monoclonal Antibody [clone: 88506]

Supplier: R & D Systems
MAB542
220021-167EA 772.25 USD
220021-167
Anti-EPHB1 Mouse Monoclonal Antibody [clone: 88506]
Antibodies
Type: Primary
Antigen: EPHB1
Clonality: Monoclonal
Clone: 88506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR