Anti-DPP9 Mouse Monoclonal Antibody [clone: 757004]

Supplier: R & D Systems
MAB5419
102886-012EA 594.37 USD
102886-012
Anti-DPP9 Mouse Monoclonal Antibody [clone: 757004]
Antibodies
Simple Western

Type: Primary
Antigen: DPP9
Clonality: Monoclonal
Clone: 757004
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR