Anti-VIPR2 Mouse Monoclonal Antibody [clone: 476031]

Supplier: R & D Systems
MAB5416
220021-165EA 577.02 USD
220021-165
Anti-VIPR2 Mouse Monoclonal Antibody [clone: 476031]
Antibodies
Type: Primary
Antigen: VIPR2
Clonality: Monoclonal
Clone: 476031
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR