Anti-PPIB Mouse Monoclonal Antibody [clone: 549205]

Supplier: R & D Systems
MAB5410
220021-164EA 633.42 USD
220021-164
Anti-PPIB Mouse Monoclonal Antibody [clone: 549205]
Antibodies
Type: Primary
Antigen: PPIB
Clonality: Monoclonal
Clone: 549205
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR