Anti-EPHA5 Mouse Monoclonal Antibody [clone: 86731]

Supplier: R & D Systems
MAB541
220021-163EA 763.57 USD
220021-163
Anti-EPHA5 Mouse Monoclonal Antibody [clone: 86731]
Antibodies
Type: Primary
Antigen: EPHA5
Clonality: Monoclonal
Clone: 86731
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR