Anti-DYRK3 Mouse Monoclonal Antibody [clone: 563518]

Supplier: R & D Systems
MAB5409
102886-010EA 577.02 USD
102886-010
Anti-DYRK3 Mouse Monoclonal Antibody [clone: 563518]
Antibodies
Type: Primary
Antigen: DYRK3
Clonality: Monoclonal
Clone: 563518
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR