Anti-CCL20 Mouse Monoclonal Antibody [clone: 83225]

Supplier: R & D Systems
MAB540
220021-162EA 772.25 USD
220021-162
Anti-CCL20 Mouse Monoclonal Antibody [clone: 83225]
Antibodies
Type: Primary
Antigen: CCL20
Clonality: Monoclonal
Clone: 83225
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR