Anti-INPPL1 Mouse Monoclonal Antibody [clone: 737935]

Supplier: R & D Systems
MAB5389
102886-006EA 566.89 USD
102886-006
Anti-INPPL1 Mouse Monoclonal Antibody [clone: 737935]
Antibodies
Type: Primary
Antigen: INPPL1
Clonality: Monoclonal
Clone: 737935
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR