Anti-PNPLA2 Mouse Monoclonal Antibody [clone: 494702]

Supplier: R & D Systems
MAB5387
220021-159EA 577.02 USD
220021-159
Anti-PNPLA2 Mouse Monoclonal Antibody [clone: 494702]
Antibodies
Type: Primary
Antigen: PNPLA2
Clonality: Monoclonal
Clone: 494702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR