Anti-TBX21 Mouse Monoclonal Antibody [clone: 525803]

Supplier: R & D Systems
MAB5385
220021-158EA 594.37 USD
220021-158
Anti-TBX21 Mouse Monoclonal Antibody [clone: 525803]
Antibodies
Type: Primary
Antigen: TBX21
Clonality: Monoclonal
Clone: 525803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR