Anti-MAG Mouse Monoclonal Antibody [clone: 118412]

Supplier: R & D Systems
MAB5381
220021-157EA 763.57 USD
220021-157
Anti-MAG Mouse Monoclonal Antibody [clone: 118412]
Antibodies
Type: Primary
Antigen: MAG
Clonality: Monoclonal
Clone: 118412
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR