Anti-ADAM32 Mouse Monoclonal Antibody [clone: 53922]

Supplier: R & D Systems
MAB5380
220021-156EA 561.11 USD
220021-156
Anti-ADAM32 Mouse Monoclonal Antibody [clone: 53922]
Antibodies
Type: Primary
Antigen: ADAM32
Clonality: Monoclonal
Clone: 53922
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR