Anti-MAG Mouse Monoclonal Antibody [clone: 118423]

Supplier: R & D Systems
MAB538
220021-155EA 763.57 USD
220021-155
Anti-MAG Mouse Monoclonal Antibody [clone: 118423]
Antibodies
Type: Primary
Antigen: MAG
Clonality: Monoclonal
Clone: 118423
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR