Anti-NCSTN Mouse Monoclonal Antibody [clone: 716918]

Supplier: R & D Systems
MAB5378
102886-002EA 561.11 USD
102886-002
Anti-NCSTN Mouse Monoclonal Antibody [clone: 716918]
Antibodies
Type: Primary
Antigen: NCSTN
Clonality: Monoclonal
Clone: 716918
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR