Anti-CD55 Rat Monoclonal Antibody [clone: 583905]

Supplier: R & D Systems
MAB5376
102886-000EA 594.37 USD
102886-000
Anti-CD55 Rat Monoclonal Antibody [clone: 583905]
Antibodies
Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: 583905
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR