Anti-PLA2G2A Mouse Monoclonal Antibody [clone: 620501]

Supplier: R & D Systems
MAB5374
102885-998EA 594.37 USD
102885-998
Anti-PLA2G2A Mouse Monoclonal Antibody [clone: 620501]
Antibodies
Type: Primary
Antigen: PLA2G2A
Clonality: Monoclonal
Clone: 620501
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR