Anti-CX3CL1 Mouse Monoclonal Antibody [clone: 96834]

Supplier: R & D Systems
MAB537
220021-154EA 763.57 USD
220021-154
Anti-CX3CL1 Mouse Monoclonal Antibody [clone: 96834]
Antibodies
Type: Primary
Antigen: CX3CL1
Clonality: Monoclonal
Clone: 96834
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR