Anti-MCSF Mouse Monoclonal Antibody [clone: 427406]

Supplier: R & D Systems
MAB5364
220021-152EA 577.02 USD
220021-152
Anti-MCSF Mouse Monoclonal Antibody [clone: 427406]
Antibodies
Type: Primary
Antigen: MCSF
Clonality: Monoclonal
Clone: 427406
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR