Anti-ITLNB Rat Monoclonal Antibody [clone: 747421]

Supplier: R & D Systems
MAB5358
102885-994EA 561.11 USD
102885-994
Anti-ITLNB Rat Monoclonal Antibody [clone: 747421]
Antibodies
Type: Primary
Antigen: ITLNB
Clonality: Monoclonal
Clone: 747421
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR