Anti-CHST4 Mouse Monoclonal Antibody [clone: 595223]

Supplier: R & D Systems
MAB5357
102885-992EA 577.02 USD
102885-992
Anti-CHST4 Mouse Monoclonal Antibody [clone: 595223]
Antibodies
Type: Primary
Antigen: CHST4
Clonality: Monoclonal
Clone: 595223
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR