Anti-SMPD1 Mouse Monoclonal Antibody [clone: 563418]

Supplier: R & D Systems
MAB5348
102885-990EA 600.15 USD
102885-990
Anti-SMPD1 Mouse Monoclonal Antibody [clone: 563418]
Antibodies
Type: Primary
Antigen: SMPD1
Clonality: Monoclonal
Clone: 563418
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR