Anti-PRKCA Rat Monoclonal Antibody [clone: 421810]

Supplier: R & D Systems
MAB5340
220021-149EA 577.02 USD
220021-149
Anti-PRKCA Rat Monoclonal Antibody [clone: 421810]
Antibodies
Type: Primary
Antigen: PRKCA
Clonality: Monoclonal
Clone: 421810
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR