Anti-CCL28 Rat Monoclonal Antibody [clone: 134315]

Supplier: R & D Systems
MAB5331
220021-148EA 763.57 USD
220021-148
Anti-CCL28 Rat Monoclonal Antibody [clone: 134315]
Antibodies
Type: Primary
Antigen: CCL28
Clonality: Monoclonal
Clone: 134315
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR