Anti-TNFRSF26 Rat Monoclonal Antibody [clone: 509027]

Supplier: R & D Systems
MAB5330
220021-147EA 561.11 USD
220021-147
Anti-TNFRSF26 Rat Monoclonal Antibody [clone: 509027]
Antibodies
Type: Primary
Antigen: TNFRSF26
Clonality: Monoclonal
Clone: 509027
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR