Anti-PLXNB2 Mouse Monoclonal Antibody [clone: 537219]

Supplier: R & D Systems
MAB53292
102885-986EA 577.02 USD
102885-986
Anti-PLXNB2 Mouse Monoclonal Antibody [clone: 537219]
Antibodies
Type: Primary
Antigen: PLXNB2
Clonality: Monoclonal
Clone: 537219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR