Anti-TGFBR2 Rat Monoclonal Antibody [clone: 129502]

Supplier: R & D Systems
MAB532
220021-144EA 763.57 USD
220021-144
Anti-TGFBR2 Rat Monoclonal Antibody [clone: 129502]
Antibodies
Type: Primary
Antigen: TGFBR2
Clonality: Monoclonal
Clone: 129502
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR