Anti-NOTCH1 Mouse Monoclonal Antibody [clone: 527425]

Supplier: R & D Systems
MAB5317-100 MAB5317-500
102885-984EA 1459.17 USD
102885-984 220021-142
Anti-NOTCH1 Mouse Monoclonal Antibody [clone: 527425]
Antibodies
Type: Primary
Antigen: NOTCH1
Clonality: Monoclonal
Clone: 527425
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR