Anti-CHST1 Mouse Monoclonal Antibody [clone: 543128]

Supplier: R & D Systems
MAB5316
102885-982EA 561.11 USD
102885-982
Anti-CHST1 Mouse Monoclonal Antibody [clone: 543128]
Antibodies
Type: Primary
Antigen: CHST1
Clonality: Monoclonal
Clone: 543128
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR