Anti-PIR-A/B Rat Monoclonal Antibody [clone: 404127]

Supplier: R & D Systems
MAB5308
220021-140EA 594.37 USD
220021-140
Anti-PIR-A/B Rat Monoclonal Antibody [clone: 404127]
Antibodies
Type: Primary
Antigen: PIR-A/B
Clonality: Monoclonal
Clone: 404127
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR