Anti-MAP3K7 Mouse Monoclonal Antibody [clone: 491840]

Supplier: R & D Systems
MAB5307
220021-139EA 594.37 USD
220021-139
Anti-MAP3K7 Mouse Monoclonal Antibody [clone: 491840]
Antibodies
Type: Primary
Antigen: MAP3K7
Clonality: Monoclonal
Clone: 491840
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR