Anti-NFE2L1 Mouse Monoclonal Antibody [clone: 492413]

Supplier: R & D Systems
MAB5306
220021-138EA 584.25 USD
220021-138
Anti-NFE2L1 Mouse Monoclonal Antibody [clone: 492413]
Antibodies
Type: Primary
Antigen Symbol: NFE2L1
Antigen Name: nuclear factor, erythroid 2-like 1
Antigen Synonyms: LCR-F1, NRF1, TCF11
Clonality: Monoclonal
Clone: 492413
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR