Anti-IL4RA Rat Monoclonal Antibody [clone: 129801]

Supplier: R & D Systems
MAB530
220021-137EA 772.25 USD
220021-137
Anti-IL4RA Rat Monoclonal Antibody [clone: 129801]
Antibodies
Type: Primary
Antigen: IL4RA
Clonality: Monoclonal
Clone: 129801
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR