Anti-POT1 Mouse Monoclonal Antibody [clone: 688923]

Supplier: R & D Systems
MAB5299
102885-980EA 577.02 USD
102885-980
Anti-POT1 Mouse Monoclonal Antibody [clone: 688923]
Antibodies
Type: Primary
Antigen: POT1
Clonality: Monoclonal
Clone: 688923
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR