Anti-BECN1 Mouse Monoclonal Antibody [clone: 669922]

Supplier: R & D Systems
MAB5295
102885-944EA 594.37 USD
102885-944
Anti-BECN1 Mouse Monoclonal Antibody [clone: 669922]
Antibodies
Simple Western

Type: Primary
Antigen: BECN1
Clonality: Monoclonal
Clone: 669922
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR