Anti-CCL17 Rat Monoclonal Antibody [clone: 110946]

Supplier: R & D Systems
MAB5291
220021-136EA 763.57 USD
220021-136
Anti-CCL17 Rat Monoclonal Antibody [clone: 110946]
Antibodies
Type: Primary
Antigen: CCL17
Clonality: Monoclonal
Clone: 110946
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR