Anti-DAPK3 Mouse Monoclonal Antibody [clone: 729830]

Supplier: R & D Systems
MAB5290
102885-940EA 577.02 USD
102885-940
Anti-DAPK3 Mouse Monoclonal Antibody [clone: 729830]
Antibodies
Type: Primary
Antigen: DAPK3
Clonality: Monoclonal
Clone: 729830
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR