Anti-STAT4 Mouse Monoclonal Antibody [clone: 513710]

Supplier: R & D Systems
MAB5287
220021-134EA 594.37 USD
220021-134
Anti-STAT4 Mouse Monoclonal Antibody [clone: 513710]
Antibodies
Type: Primary
Antigen: STAT4
Clonality: Monoclonal
Clone: 513710
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR